Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GMFG
Entrez Gene ID
9535
Gene Name
glia maturation factor gamma
Gene Aliases
GMF-GAMMA
Chromosome Location
Chr.19: 39328357 - 39336086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004877.3 NP_004868.1 1 - 2 111 93
GenBank mRNA BC022207.1 - 1 - 2 31 93
BC080180.1 - 1 - 2 31 93
BC093799.1 - 1 - 2 64 93
BC101818.1 - 1 - 2 64 93
BC143543.1 - 1 - 2 64 93
BC143546.1 - 1 - 2 63 93
BC143548.1 - 1 - 2 62 93
BF974742.1 - 1 - 2 112 93
EB387540.1 - 1 - 2 210 93

View Full Product Details