The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283050.1 | NP_001269979.1 | 3 - 4 | 594 | 134 |
NM_001283052.1 | NP_001269981.1 | 3 - 4 | 442 | 134 | |
NM_015259.5 | NP_056074.1 | 3 - 4 | 594 | 134 | |
XM_011529514.2 | XP_011527816.1 | 3 - 4 | 1037 | 134 | |
XM_011529515.2 | XP_011527817.1 | 3 - 4 | 1037 | 134 | |
XM_011529516.2 | XP_011527818.1 | 2 - 3 | 429 | 134 | |
XM_017028310.1 | XP_016883799.1 | 3 - 4 | 884 | 134 | |
GenBank mRNA | AB014553.1 | - | 3 - 4 | 535 | 134 |
AF199028.1 | - | 3 - 4 | 430 | 134 | |
AF216749.1 | - | 2 - 3 | 407 | 134 | |
AF289028.1 | - | 3 - 4 | 531 | 134 | |
AK290192.1 | - | 3 - 4 | 556 | 134 | |
AK294031.1 | - | 3 - 4 | 404 | 134 | |
AK311496.1 | - | 3 - 4 | 556 | 134 | |
AK314369.1 | - | 3 - 4 | 556 | 134 | |
AK316143.1 | - | 1 - 2 | 269 | 134 | |
AK316176.1 | - | 1 - 2 | 237 | 134 | |
BC064637.1 | - | 3 - 4 | 515 | 134 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006723899.2 | XP_006723962.1 | 3 - 4 | 1043 | 134 |
XM_006723900.2 | XP_006723963.1 | 3 - 4 | 1043 | 134 | |
XM_011546078.2 | XP_011544380.1 | 3 - 4 | 1043 | 134 | |
XM_011546079.1 | XP_011544381.1 | 3 - 4 | 1043 | 134 |