The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307943.1 | NP_001294872.1 | 12 - 13 | 1666 | 76 |
NM_018259.5 | NP_060729.2 | 12 - 13 | 1666 | 76 | |
XM_005253016.1 | XP_005253073.1 | 12 - 13 | 1666 | 76 | |
XM_006718263.1 | XP_006718326.1 | 10 - 11 | 1349 | 76 | |
XM_011520218.2 | XP_011518520.1 | 12 - 13 | 1666 | 76 | |
XM_017018010.1 | XP_016873499.1 | 12 - 13 | 1666 | 76 | |
GenBank mRNA | AK001540.1 | - | 7 - 8 | 901 | 76 |
AK001752.1 | - | 12 - 13 | 1619 | 76 | |
AK074253.1 | - | 2 - 3 | 183 | 76 | |
BC017238.2 | - | 12 - 13 | 1615 | 76 | |
BC033000.1 | - | 12 - 13 | 1612 | 76 | |
BX640972.1 | - | 7 - 8 | 1506 | 76 |