Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SCAP
Entrez Gene ID
22937
Gene Name
SREBF chaperone
Gene Aliases
-
Chromosome Location
Chr.3: 47413685 - 47477126 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012235.3 NP_036367.2 4 - 5 667 62
XM_005264967.1 XP_005265024.1 4 - 5 594 62
XM_011533501.1 XP_011531803.1 4 - 5 694 62
XM_017005918.1 XP_016861407.1 4 - 5 681 62
XM_017005919.1 XP_016861408.1 4 - 5 614 62
XM_017005920.1 XP_016861409.1 4 - 5 614 62
GenBank mRNA D83782.2 - 4 - 5 653 62

View Full Product Details