Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRHL2
Entrez Gene ID
79977
Gene Name
grainyhead like transcription factor 2
Gene Aliases
BOM, DFNA28, ECTDS, TFCP2L3
Chromosome Location
Chr.8: 101492440 - 101678105 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024915.3 NP_079191.2 5 - 6 1067 64
XM_011517305.2 XP_011515607.1 5 - 6 1098 64
XM_011517306.2 XP_011515608.1 5 - 6 835 64
XM_011517307.2 XP_011515609.1 5 - 6 1065 64
GenBank mRNA AK023844.1 - 5 - 6 749 64
AK292853.1 - 5 - 6 749 64
AK296817.1 - 5 - 6 1066 64
BC069618.1 - 5 - 6 749 64
BC069633.1 - 5 - 6 809 64
BC069638.1 - 5 - 6 749 64
BC129822.1 - 5 - 6 749 64
BC129823.1 - 5 - 6 749 64

View Full Product Details