The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024915.3 | NP_079191.2 | 6 - 7 | 1218 | 106 |
XM_011517305.2 | XP_011515607.1 | 6 - 7 | 1249 | 106 | |
XM_011517306.2 | XP_011515608.1 | 6 - 7 | 986 | 106 | |
XM_011517307.2 | XP_011515609.1 | 6 - 7 | 1216 | 106 | |
GenBank mRNA | AK023844.1 | - | 6 - 7 | 900 | 106 |
AK292853.1 | - | 6 - 7 | 900 | 106 | |
BC069618.1 | - | 6 - 7 | 900 | 106 | |
BC069633.1 | - | 6 - 7 | 960 | 106 | |
BC069638.1 | - | 6 - 7 | 900 | 106 | |
BC129822.1 | - | 6 - 7 | 900 | 106 | |
BC129823.1 | - | 6 - 7 | 900 | 106 |