Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STC1
Entrez Gene ID
6781
Gene Name
stanniocalcin 1
Gene Aliases
STC
Chromosome Location
Chr.8: 23841921 - 23854807 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003155.2 NP_003146.1 2 - 3 549 69
GenBank mRNA AK297734.1 - 3 - 4 396 69
AK309366.1 - 2 - 3 413 69
AK312589.1 - 2 - 3 548 69
BC029044.1 - 2 - 3 527 69
KP455655.1 - 2 - 3 212 69
KP455656.1 - 2 - 3 212 69
KP455657.1 - 2 - 3 212 69
KP455658.1 - 2 - 3 213 69
KP455659.1 - 2 - 3 212 69
U25997.1 - 2 - 3 549 69
U46768.1 - 2 - 3 299 69

View Full Product Details