The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002291.2 | NP_002282.2 | 12 - 13 | 1815 | 73 |
XM_017012201.1 | XP_016867690.1 | 12 - 13 | 2283 | 73 | |
XM_017012202.1 | XP_016867691.1 | 12 - 13 | 2283 | 73 | |
GenBank mRNA | AK307611.1 | - | 12 - 13 | 1635 | 73 |
AK309921.1 | - | 13 - 14 | 1891 | 73 | |
BC113455.1 | - | 11 - 12 | 1523 | 73 | |
BX648251.1 | - | 11 - 12 | 1744 | 73 | |
M61916.1 | - | 12 - 13 | 1597 | 73 |