Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSME4
Entrez Gene ID
23198
Gene Name
proteasome activator subunit 4
Gene Aliases
PA200
Chromosome Location
Chr.2: 53864067 - 53970840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014614.2 NP_055429.2 31 - 32 3656 159
XM_006711969.1 XP_006712032.1 30 - 31 3233 159
XM_011532705.2 XP_011531007.1 32 - 33 3680 159
XM_011532706.2 XP_011531008.1 32 - 33 3680 159
XM_011532707.2 XP_011531009.1 31 - 32 3614 159
GenBank mRNA AK026085.1 - 5 - 6 653 159
AY894754.1 - 30 - 31 3505 159
AY894755.1 - 12 - 13 1344 159
BC112169.1 - 30 - 31 3292 159
BC113668.1 - 30 - 31 3292 159
BC143739.1 - 30 - 31 3292 159
D38521.1 - 31 - 32 3466 159

View Full Product Details