Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSME4
Entrez Gene ID
23198
Gene Name
proteasome activator subunit 4
Gene Aliases
PA200
Chromosome Location
Chr.2: 53864067 - 53970840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014614.2 NP_055429.2 37 - 38 4350 142
XM_006711969.1 XP_006712032.1 36 - 37 3927 142
XM_011532705.2 XP_011531007.1 38 - 39 4374 142
XM_011532706.2 XP_011531008.1 38 - 39 4374 142
XM_011532707.2 XP_011531009.1 37 - 38 4308 142
GenBank mRNA AK025517.1 - 6 - 7 694 142
AK026085.1 - 11 - 12 1347 142
AY894754.1 - 36 - 37 4199 142
AY894755.1 - 18 - 19 2038 142
BC043602.1 - 2 - 3 144 142
BC112169.1 - 36 - 37 3986 142
BC113668.1 - 36 - 37 3986 142
BC143739.1 - 36 - 37 3986 142
D38521.1 - 37 - 38 4160 142

View Full Product Details