The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286791.1 | NP_001273720.1 | 4 - 5 | 619 | 76 |
NM_025087.2 | NP_079363.2 | 4 - 5 | 700 | 76 | |
XM_011513755.1 | XP_011512057.1 | 4 - 5 | 701 | 76 | |
XM_011513756.2 | XP_011512058.1 | 4 - 5 | 732 | 76 | |
XM_011513757.2 | XP_011512059.1 | 4 - 5 | 758 | 76 | |
XM_011513758.1 | XP_011512060.1 | 4 - 5 | 701 | 76 | |
GenBank mRNA | AK025164.1 | - | 4 - 5 | 700 | 76 |
AK300495.1 | - | 4 - 5 | 622 | 76 | |
AK316307.1 | - | 4 - 5 | 592 | 76 | |
BC137387.1 | - | 4 - 5 | 683 | 76 | |
BC137388.1 | - | 4 - 5 | 683 | 76 |