The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318792.1 | NP_001305721.1 | 14 - 15 | 1444 | 99 |
NM_018426.2 | NP_060896.1 | 14 - 15 | 1410 | 99 | |
XM_005249211.3 | XP_005249268.1 | 16 - 17 | 1790 | 99 | |
XM_005249213.4 | XP_005249270.1 | 15 - 16 | 1717 | 99 | |
XM_005249214.4 | XP_005249271.1 | 13 - 14 | 1304 | 99 | |
XM_005249215.4 | XP_005249272.1 | 14 - 15 | 1609 | 99 | |
XM_005249217.1 | XP_005249274.1 | 14 - 15 | 1482 | 99 | |
XM_006715135.1 | XP_006715198.1 | 14 - 15 | 1466 | 99 | |
XM_017010999.1 | XP_016866488.1 | 16 - 17 | 1667 | 99 | |
XM_017011000.1 | XP_016866489.1 | 15 - 16 | 1648 | 99 | |
XM_017011001.1 | XP_016866490.1 | 13 - 14 | 1348 | 99 | |
XM_017011002.1 | XP_016866491.1 | 13 - 14 | 1451 | 99 | |
GenBank mRNA | AL834238.1 | - | 7 - 8 | 708 | 99 |
BC022095.1 | - | 11 - 12 | 1047 | 99 | |
BC062989.1 | - | 2 - 3 | 236 | 99 | |
BC136769.1 | - | 13 - 14 | 1286 | 99 | |
CR627460.1 | - | 14 - 15 | 1447 | 99 |