The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318792.1 | NP_001305721.1 | 7 - 8 | 733 | 65 |
NM_018426.2 | NP_060896.1 | 7 - 8 | 699 | 65 | |
XM_005249211.3 | XP_005249268.1 | 9 - 10 | 1079 | 65 | |
XM_005249213.4 | XP_005249270.1 | 8 - 9 | 1006 | 65 | |
XM_005249214.4 | XP_005249271.1 | 6 - 7 | 593 | 65 | |
XM_005249215.4 | XP_005249272.1 | 7 - 8 | 898 | 65 | |
XM_005249217.1 | XP_005249274.1 | 7 - 8 | 771 | 65 | |
XM_006715135.1 | XP_006715198.1 | 7 - 8 | 755 | 65 | |
XM_017010999.1 | XP_016866488.1 | 9 - 10 | 956 | 65 | |
XM_017011000.1 | XP_016866489.1 | 8 - 9 | 937 | 65 | |
XM_017011001.1 | XP_016866490.1 | 6 - 7 | 637 | 65 | |
XM_017011002.1 | XP_016866491.1 | 6 - 7 | 740 | 65 | |
GenBank mRNA | AK093629.1 | - | 7 - 8 | 755 | 65 |
AK309791.1 | - | 5 - 6 | 602 | 65 | |
BC022095.1 | - | 4 - 5 | 336 | 65 | |
BC136769.1 | - | 6 - 7 | 575 | 65 | |
CR627460.1 | - | 7 - 8 | 736 | 65 |