The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001126240.2 | NP_001119712.1 | 8 - 9 | 1284 | 108 |
NM_001126241.2 | NP_001119713.1 | 8 - 9 | 1284 | 108 | |
NM_001204184.1 | NP_001191113.1 | 10 - 11 | 1307 | 108 | |
NM_001204192.1 | NP_001191121.1 | 8 - 9 | 1117 | 108 | |
NM_005427.3 | NP_005418.1 | 10 - 11 | 1307 | 108 | |
GenBank mRNA | AB055065.1 | - | 8 - 9 | 1284 | 108 |
AB055066.1 | - | 8 - 9 | 1284 | 108 | |
AK302118.1 | - | 8 - 9 | 1105 | 108 | |
AK304784.1 | - | 8 - 9 | 1234 | 108 | |
AY040827.1 | - | 8 - 9 | 1284 | 108 | |
AY040828.1 | - | 8 - 9 | 1284 | 108 | |
BC117251.1 | - | 10 - 11 | 1307 | 108 | |
BC117253.1 | - | 10 - 11 | 1307 | 108 | |
Y11416.1 | - | 10 - 11 | 1307 | 108 |