The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000030.2 | NP_000021.1 | 9 - 10 | 1060 | 64 |
GenBank mRNA | AB292648.1 | - | 9 - 10 | 961 | 64 |
AF191687.1 | - | 1 - 2 | 717 | 64 | |
AF348451.1 | - | 9 - 10 | 851 | 64 | |
AK292754.1 | - | 9 - 10 | 981 | 64 | |
AK315331.1 | - | 9 - 10 | 1008 | 64 | |
BC132819.1 | - | 9 - 10 | 1057 | 64 | |
D13368.1 | - | 9 - 10 | 1047 | 64 | |
X53414.1 | - | 9 - 10 | 1061 | 64 | |
X56092.1 | - | 9 - 10 | 960 | 64 |