The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178129.1 | NP_001171600.1 | 10 - 11 | 1127 | 84 |
NM_012431.2 | NP_036563.1 | 10 - 11 | 1763 | 84 | |
GenBank mRNA | AB002329.1 | - | 10 - 11 | 1608 | 84 |
AK297209.1 | - | 10 - 11 | 1443 | 84 | |
AK303925.1 | - | 10 - 11 | 1128 | 84 | |
BC140706.1 | - | 10 - 11 | 1298 | 84 | |
BC144338.1 | - | 10 - 11 | 1298 | 84 | |
BC152458.1 | - | 10 - 11 | 1620 | 84 |