The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178129.1 | NP_001171600.1 | 13 - 14 | 1484 | 132 |
NM_012431.2 | NP_036563.1 | 13 - 14 | 2120 | 132 | |
GenBank mRNA | AK297209.1 | - | 13 - 14 | 1800 | 132 |
AK303925.1 | - | 13 - 14 | 1485 | 132 | |
BC140706.1 | - | 13 - 14 | 1655 | 132 | |
BC144338.1 | - | 13 - 14 | 1655 | 132 |