Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPN
Entrez Gene ID
3249
Gene Name
hepsin
Gene Aliases
TMPRSS1
Chromosome Location
Chr.19: 35040506 - 35066573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002151.2 NP_002142.1 6 - 7 536 89
NM_182983.2 NP_892028.1 5 - 6 1116 89
XM_005258838.4 XP_005258895.2 5 - 6 614 89
XM_006723181.3 XP_006723244.2 5 - 6 490 89
XM_017026731.1 XP_016882220.1 5 - 6 789 89
XM_017026732.1 XP_016882221.1 4 - 5 351 89
GenBank mRNA AK293622.1 - 4 - 5 370 89
AK315655.1 - 5 - 6 469 89
BC025716.1 - 5 - 6 488 89
EU644753.1 - 4 - 5 291 89
M18930.1 - 6 - 7 536 89
X07732.1 - 5 - 6 1116 89

View Full Product Details