Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HPN
Entrez Gene ID
3249
Gene Name
hepsin
Gene Aliases
TMPRSS1
Chromosome Location
Chr.19: 35040506 - 35066573 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002151.2 NP_002142.1 9 - 10 868 81
NM_182983.2 NP_892028.1 8 - 9 1448 81
XM_005258838.4 XP_005258895.2 8 - 9 946 81
XM_006723181.3 XP_006723244.2 8 - 9 822 81
XM_017026731.1 XP_016882220.1 8 - 9 1121 81
XM_017026732.1 XP_016882221.1 7 - 8 683 81
GenBank mRNA AK095160.1 - 2 - 3 1044 81
AK293622.1 - 7 - 8 702 81
AK315655.1 - 8 - 9 801 81
BC025716.1 - 8 - 9 820 81
EU644753.1 - 7 - 8 623 81
M18930.1 - 9 - 10 868 81
X07002.1 - 3 - 4 284 81
X07732.1 - 8 - 9 1448 81

View Full Product Details