The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320289.1 | NP_001307218.1 | 8 - 9 | 1291 | 109 |
NM_001320290.1 | NP_001307219.1 | 8 - 9 | 1028 | 109 | |
NM_178033.1 | NP_828847.1 | 8 - 9 | 1325 | 109 | |
XM_017000973.1 | XP_016856462.1 | 8 - 9 | 1356 | 109 | |
GenBank mRNA | AK091806.1 | - | 8 - 9 | 1029 | 109 |
AK098065.1 | - | 8 - 9 | 1318 | 109 | |
AK131355.1 | - | 8 - 9 | 1291 | 109 | |
AM040940.1 | - | 8 - 9 | 1162 | 109 | |
AY358537.1 | - | 8 - 9 | 1308 | 109 | |
BC028102.1 | - | 8 - 9 | 1325 | 109 |