The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128852.1 | NP_001122324.1 | 11 - 12 | 1640 | 76 |
NM_001128853.1 | NP_001122325.1 | 11 - 12 | 1643 | 76 | |
NM_001128854.1 | NP_001122326.1 | 11 - 12 | 1640 | 76 | |
NM_015908.5 | NP_056992.4 | 11 - 12 | 1643 | 76 | |
XM_005250405.2 | XP_005250462.1 | 11 - 12 | 1500 | 76 | |
XM_005250406.2 | XP_005250463.1 | 11 - 12 | 1497 | 76 | |
XM_005250407.2 | XP_005250464.1 | 11 - 12 | 1500 | 76 | |
XM_005250408.2 | XP_005250465.1 | 11 - 12 | 1497 | 76 | |
XM_017012290.1 | XP_016867779.1 | 12 - 13 | 1825 | 76 | |
XM_017012291.1 | XP_016867780.1 | 12 - 13 | 1822 | 76 | |
GenBank mRNA | AF082871.1 | - | 1 - 2 | 51 | 76 |
AF248955.1 | - | 11 - 12 | 1606 | 76 | |
AK296131.1 | - | 5 - 6 | 609 | 76 | |
AK302820.1 | - | 11 - 12 | 1532 | 76 | |
AL096723.1 | - | 11 - 12 | 1595 | 76 | |
BC000082.2 | - | 2 - 3 | 164 | 76 | |
BC069249.1 | - | 9 - 10 | 1184 | 76 | |
BC109117.1 | - | 11 - 12 | 1534 | 76 |