The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003227.3 | NP_003218.2 | 1 - 2 | 73 | 71 |
XM_005250553.4 | XP_005250610.1 | 2 - 3 | 379 | 71 | |
XM_017012573.1 | XP_016868062.1 | 2 - 3 | 181 | 71 | |
GenBank mRNA | AF067864.1 | - | 1 - 2 | 59 | 71 |
AK022002.1 | - | 2 - 3 | 190 | 71 | |
BC142630.1 | - | 1 - 2 | 59 | 71 | |
BC150192.1 | - | 1 - 2 | 37 | 71 | |
DR004213.1 | - | 1 - 2 | 217 | 71 |