Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTK2
Entrez Gene ID
5747
Gene Name
protein tyrosine kinase 2
Gene Aliases
FADK, FAK, FAK1, FRNK, PPP1R71, p125FAK, pp125FAK
Chromosome Location
Chr.8: 140658382 - 141001313 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199649.1 NP_001186578.1 16 - 17 1640 73
NM_001316342.1 NP_001303271.1 16 - 17 1423 73
NM_005607.4 NP_005598.3 16 - 17 1629 73
NM_153831.3 NP_722560.1 16 - 17 1640 73
XM_005251003.2 XP_005251060.1 18 - 19 1804 73
XM_005251012.2 XP_005251069.1 18 - 19 1646 73
XM_005251013.2 XP_005251070.1 17 - 18 1465 73
XM_006716608.2 XP_006716671.1 19 - 20 1735 73
XM_006716609.2 XP_006716672.1 18 - 19 1568 73
XM_006716610.2 XP_006716673.1 18 - 19 1560 73
XM_011517194.2 XP_011515496.1 19 - 20 1704 73
XM_017013651.1 XP_016869140.1 17 - 18 1485 73
XM_017013652.1 XP_016869141.1 17 - 18 1485 73
XM_017013653.1 XP_016869142.1 17 - 18 1720 73
XM_017013654.1 XP_016869143.1 16 - 17 1702 73
XM_017013655.1 XP_016869144.1 17 - 18 1696 73
XM_017013656.1 XP_016869145.1 16 - 17 1702 73
XM_017013657.1 XP_016869146.1 15 - 16 1681 73
XM_017013658.1 XP_016869147.1 18 - 19 1607 73
XM_017013659.1 XP_016869148.1 14 - 15 1362 73
XM_017013660.1 XP_016869149.1 18 - 19 1544 73
XM_017013661.1 XP_016869150.1 17 - 18 1562 73
XM_017013662.1 XP_016869151.1 17 - 18 1526 73
XM_017013663.1 XP_016869152.1 16 - 17 1457 73
XM_017013664.1 XP_016869153.1 17 - 18 1630 73
XM_017013665.1 XP_016869154.1 16 - 17 1454 73
XM_017013666.1 XP_016869155.1 14 - 15 1573 73
XM_017013667.1 XP_016869156.1 18 - 19 1520 73
XM_017013668.1 XP_016869157.1 17 - 18 1538 73
XM_017013669.1 XP_016869158.1 17 - 18 1526 73
XM_017013670.1 XP_016869159.1 17 - 18 1523 73
XM_017013671.1 XP_016869160.1 16 - 17 1505 73
XM_017013672.1 XP_016869161.1 15 - 16 1436 73
XM_017013673.1 XP_016869162.1 15 - 16 1436 73
XM_017013674.1 XP_016869163.1 18 - 19 1651 73
XM_017013675.1 XP_016869164.1 17 - 18 1475 73
XM_017013676.1 XP_016869165.1 14 - 15 1362 73
XM_017013677.1 XP_016869166.1 17 - 18 1436 73
XM_017013678.1 XP_016869167.1 14 - 15 1328 73
XM_017013679.1 XP_016869168.1 16 - 17 1367 73
XM_017013680.1 XP_016869169.1 18 - 19 2620 73
XM_017013681.1 XP_016869170.1 18 - 19 1568 73
XM_017013682.1 XP_016869171.1 16 - 17 1444 73
XM_017013683.1 XP_016869172.1 17 - 18 1484 73
XM_017013684.1 XP_016869173.1 16 - 17 1466 73
XM_017013685.1 XP_016869174.1 16 - 17 1407 73
XM_017013686.1 XP_016869175.1 14 - 15 1342 73
XM_017013687.1 XP_016869176.1 17 - 18 1484 73
XM_017013688.1 XP_016869177.1 16 - 17 1376 73
XM_017013689.1 XP_016869178.1 15 - 16 1241 73
XM_017013690.1 XP_016869179.1 14 - 15 1197 73
GenBank mRNA AB209071.1 - 16 - 17 1329 73
AB209083.1 - 19 - 20 1350 73
AK094999.1 - 6 - 7 346 73
AK294873.1 - 7 - 8 424 73
AK304356.1 - 16 - 17 1485 73
AL832961.1 - 16 - 17 1512 73
BC028733.2 - 2 - 3 288 73
BC035404.2 - 14 - 15 1361 73
KF437463.1 - 14 - 15 1333 73
L13616.1 - 16 - 17 1562 73

View Full Product Details