Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A40
Entrez Gene ID
55972
Gene Name
solute carrier family 25 member 40
Gene Aliases
MCFP
Chromosome Location
Chr.7: 87834499 - 87876377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018843.3 NP_061331.2 6 - 7 714 139
GenBank mRNA AF125531.1 - 7 - 8 687 139
AK223020.1 - 6 - 7 668 139
AK290848.1 - 6 - 7 489 139
BC027322.1 - 5 - 6 446 139

View Full Product Details