The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018843.3 | NP_061331.2 | 8 - 9 | 1002 | 136 |
GenBank mRNA | AF125531.1 | - | 9 - 10 | 975 | 136 |
AK223020.1 | - | 8 - 9 | 956 | 136 | |
AK290848.1 | - | 8 - 9 | 777 | 136 | |
BC027322.1 | - | 7 - 8 | 734 | 136 |