Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CROT
Entrez Gene ID
54677
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
COT
Chromosome Location
Chr.7: 87345635 - 87399796 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243745.1 NP_001230674.1 3 - 4 357 155
NM_021151.3 NP_066974.2 3 - 4 357 155
XM_011516337.2 XP_011514639.1 3 - 4 357 155
XM_017012370.1 XP_016867859.1 3 - 4 295 155
GenBank mRNA AF073770.1 - 3 - 4 248 155
AF168793.1 - 3 - 4 248 155
AK313487.1 - 3 - 4 301 155
BC006015.2 - 2 - 3 227 155
BC039004.1 - 3 - 4 297 155
BC051874.1 - 3 - 4 299 155
DA518578.1 - 3 - 4 355 155
DA865171.1 - 3 - 4 357 155

View Full Product Details