The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013448.2 | NP_038476.2 | 14 - 15 | 2392 | 90 |
NM_182648.1 | NP_872589.1 | 13 - 14 | 2296 | 90 | |
XM_011536374.1 | XP_011534676.1 | 13 - 14 | 1691 | 90 | |
XM_011536376.2 | XP_011534678.2 | 4 - 5 | 519 | 90 | |
XM_017020942.1 | XP_016876431.1 | 3 - 4 | 422 | 90 | |
GenBank mRNA | AB032252.1 | - | 13 - 14 | 2293 | 90 |
AB209255.1 | - | 5 - 6 | 643 | 90 | |
AF213467.1 | - | 14 - 15 | 1990 | 90 | |
AF221130.1 | - | 13 - 14 | 1824 | 90 |