The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013448.2 | NP_038476.2 | 17 - 18 | 2804 | 112 |
NM_182648.1 | NP_872589.1 | 16 - 17 | 2708 | 112 | |
XM_011536374.1 | XP_011534676.1 | 16 - 17 | 2103 | 112 | |
XM_011536376.2 | XP_011534678.2 | 7 - 8 | 931 | 112 | |
XM_017020942.1 | XP_016876431.1 | 6 - 7 | 834 | 112 | |
GenBank mRNA | AB209255.1 | - | 8 - 9 | 1055 | 112 |
AF213467.1 | - | 17 - 18 | 2402 | 112 | |
AF221130.1 | - | 16 - 17 | 2236 | 112 |