The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013448.2 | NP_038476.2 | 3 - 4 | 966 | 138 |
NM_182648.1 | NP_872589.1 | 3 - 4 | 966 | 138 | |
GenBank mRNA | AF161435.1 | - | 3 - 4 | 652 | 138 |
AF213467.1 | - | 3 - 4 | 564 | 138 | |
AF221130.1 | - | 2 - 3 | 398 | 138 | |
BC020636.1 | - | 2 - 3 | 726 | 138 | |
BC108711.1 | - | 3 - 4 | 630 | 138 |