Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BAZ1A
Entrez Gene ID
11177
Gene Name
bromodomain adjacent to zinc finger domain 1A
Gene Aliases
ACF1, WALp1, WCRF180, hACF1
Chromosome Location
Chr.14: 34752731 - 34875647 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013448.2 NP_038476.2 6 - 7 1293 115
NM_182648.1 NP_872589.1 6 - 7 1293 115
XM_011536374.1 XP_011534676.1 5 - 6 592 115
GenBank mRNA AB032252.1 - 6 - 7 1290 115
AF161435.1 - 6 - 7 979 115
AF213467.1 - 6 - 7 891 115
AF221130.1 - 5 - 6 725 115
BC020636.1 - 5 - 6 1053 115
BC108711.1 - 6 - 7 957 115

View Full Product Details