The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006393.2 | NP_006384.1 | 10 - 11 | 1407 | 130 |
XM_005252342.4 | XP_005252399.1 | 9 - 10 | 1409 | 130 | |
XM_005252343.4 | XP_005252400.1 | 10 - 11 | 1511 | 130 | |
XM_005252344.4 | XP_005252401.1 | 10 - 11 | 1511 | 130 | |
XM_011519291.2 | XP_011517593.1 | 12 - 13 | 1403 | 130 | |
XM_017015467.1 | XP_016870956.1 | 12 - 13 | 1519 | 130 | |
XM_017015468.1 | XP_016870957.1 | 13 - 14 | 1287 | 130 | |
XM_017015469.1 | XP_016870958.1 | 10 - 11 | 1511 | 130 | |
GenBank mRNA | BC110452.1 | - | 10 - 11 | 1094 | 130 |
BC110453.1 | - | 10 - 11 | 1094 | 130 | |
Y16241.1 | - | 10 - 11 | 1407 | 130 |