Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEBL
Entrez Gene ID
10529
Gene Name
nebulette
Gene Aliases
LASP2, LNEBL
Chromosome Location
Chr.10: 20779973 - 21174187 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006393.2 NP_006384.1 24 - 25 2919 87
XM_005252342.4 XP_005252399.1 23 - 24 2921 87
XM_011519291.2 XP_011517593.1 26 - 27 2915 87
XM_017015467.1 XP_016870956.1 26 - 27 3031 87
XM_017015468.1 XP_016870957.1 27 - 28 2799 87
GenBank mRNA BC110452.1 - 24 - 25 3326 87
BC110453.1 - 24 - 25 2606 87
Y16241.1 - 24 - 25 2919 87

View Full Product Details