The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000187.3 | NP_000178.2 | 12 - 13 | 1467 | 113 |
XM_005247412.1 | XP_005247469.1 | 10 - 11 | 1242 | 113 | |
XM_017006277.1 | XP_016861766.1 | 12 - 13 | 1080 | 113 | |
GenBank mRNA | AF045167.1 | - | 12 - 13 | 1174 | 113 |
AK124291.1 | - | 7 - 8 | 1851 | 113 | |
AK290782.1 | - | 12 - 13 | 1173 | 113 | |
AK313563.1 | - | 12 - 13 | 1451 | 113 | |
BC071757.1 | - | 12 - 13 | 1105 | 113 | |
U63008.1 | - | 12 - 13 | 1136 | 113 | |
Z75048.1 | - | 12 - 13 | 1136 | 113 |