Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NT5C2
- Entrez Gene ID
-
22978
- Gene Name
- 5'-nucleotidase, cytosolic II
- Gene Aliases
-
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
- Chromosome Location
-
Chr.10: 103088017 - 103193306 on Build GRCh38
View Full Product Details