Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAN
Entrez Gene ID
8139
Gene Name
gigaxonin
Gene Aliases
GAN1, KLHL16
Chromosome Location
Chr.16: 81314941 - 81380198 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022041.3 NP_071324.1 1 - 2 316 156
GenBank mRNA AF291673.1 - 1 - 2 312 156
AK095378.1 - 1 - 2 316 156
BC044840.1 - 1 - 2 303 156
KU178409.1 - 1 - 2 168 156

View Full Product Details