The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193314.1 | NP_001180243.1 | 3 - 4 | 737 | 91 |
NM_001193315.1 | NP_001180244.1 | 3 - 4 | 337 | 91 | |
NM_001193317.1 | NP_001180246.1 | 3 - 4 | 514 | 91 | |
NM_022067.3 | NP_071350.2 | 4 - 5 | 608 | 91 | |
XM_011537066.1 | XP_011535368.1 | 3 - 4 | 328 | 91 | |
XM_017021579.1 | XP_016877068.1 | 3 - 4 | 462 | 91 | |
XM_017021580.1 | XP_016877069.1 | 3 - 4 | 328 | 91 | |
XM_017021581.1 | XP_016877070.1 | 3 - 4 | 462 | 91 | |
GenBank mRNA | AK022769.1 | - | 3 - 4 | 285 | 91 |
AK022925.1 | - | 3 - 4 | 263 | 91 | |
AK098254.1 | - | 3 - 4 | 698 | 91 | |
AK222706.1 | - | 3 - 4 | 283 | 91 | |
AK297388.1 | - | 3 - 4 | 439 | 91 | |
BC015054.1 | - | 4 - 5 | 529 | 91 | |
CR457294.1 | - | 2 - 3 | 191 | 91 | |
DA794284.1 | - | 3 - 4 | 337 | 91 |