The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003659.3 | NP_003650.1 | 13 - 14 | 1411 | 156 |
XM_011512041.2 | XP_011510343.1 | 13 - 14 | 1193 | 156 | |
XM_011512042.1 | XP_011510344.1 | 13 - 14 | 1360 | 156 | |
XM_011512043.2 | XP_011510345.1 | 9 - 10 | 1997 | 156 | |
GenBank mRNA | AK296239.1 | - | 13 - 14 | 1191 | 156 |
AK309714.1 | - | 13 - 14 | 1510 | 156 | |
AK314259.1 | - | 13 - 14 | 1381 | 156 | |
AY544121.1 | - | 13 - 14 | 1364 | 156 | |
BC141820.1 | - | 13 - 14 | 1376 | 156 | |
Y09443.1 | - | 13 - 14 | 1379 | 156 |