Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- POLL
- Entrez Gene ID
-
27343
- Gene Name
- polymerase (DNA) lambda
- Gene Aliases
-
BETAN, POLKAPPA
- Chromosome Location
-
Chr.10: 101578882 - 101588270 on Build GRCh38
View Full Product Details