Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPAST
Entrez Gene ID
6683
Gene Name
spastin
Gene Aliases
ADPSP, FSP2, SPG4
Chromosome Location
Chr.2: 32063551 - 32157637 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014946.3 NP_055761.2 8 - 9 1393 140
NM_199436.1 NP_955468.1 7 - 8 1297 140
XM_005264516.4 XP_005264573.1 8 - 9 1169 140
XM_011533067.2 XP_011531369.1 8 - 9 1453 140
XM_017004777.1 XP_016860266.1 7 - 8 1354 140
XM_017004778.1 XP_016860267.1 7 - 8 1357 140
GenBank mRNA AB029006.1 - 7 - 8 1297 140
AJ246001.1 - 8 - 9 1297 140
BC150260.1 - 7 - 8 1297 140

View Full Product Details