The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014633.4 | NP_055448.1 | 12 - 13 | 2022 | 107 |
GenBank mRNA | AK291794.1 | - | 12 - 13 | 1758 | 107 |
BC058914.1 | - | 12 - 13 | 1769 | 107 | |
D63875.1 | - | 12 - 13 | 1682 | 107 |