The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014633.4 | NP_055448.1 | 8 - 9 | 1385 | 107 |
GenBank mRNA | AK291794.1 | - | 8 - 9 | 1121 | 107 |
BC058914.1 | - | 8 - 9 | 1132 | 107 | |
D63875.1 | - | 8 - 9 | 1045 | 107 |