Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS51
Entrez Gene ID
738
Gene Name
VPS51, GARP complex subunit
Gene Aliases
ANG2, ANG3, C11orf2, C11orf3, FFR
Chromosome Location
Chr.11: 65096115 - 65111860 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013265.3 NP_037397.2 9 - 10 2226 68
NR_073519.1 9 - 10 2228 68
GenBank mRNA AF024631.2 - 9 - 10 2130 68
AL833818.1 - 8 - 9 1766 68
BC006555.2 - 8 - 9 1872 68
BC007198.1 - 9 - 10 2099 68
BC009285.2 - 5 - 6 1303 68
BC010540.1 - 9 - 10 2105 68
BC010872.2 - 9 - 10 2101 68
BC017438.1 - 9 - 10 2099 68

View Full Product Details