The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098670.1 | NP_001092140.1 | 2 - 3 | 257 | 91 |
NM_001098671.1 | NP_001092141.1 | 2 - 3 | 297 | 91 | |
NM_153819.1 | NP_722541.1 | 2 - 3 | 323 | 91 | |
XM_011544720.1 | XP_011543022.1 | 2 - 3 | 728 | 91 | |
XM_011544721.1 | XP_011543023.1 | 2 - 3 | 260 | 91 | |
XM_011544722.1 | XP_011543024.1 | 2 - 3 | 313 | 91 | |
XM_011544723.2 | XP_011543025.1 | 2 - 3 | 262 | 91 | |
XM_017017084.1 | XP_016872573.1 | 2 - 3 | 291 | 91 | |
GenBank mRNA | AF043722.1 | - | 3 - 4 | 429 | 91 |
AF043723.1 | - | 2 - 3 | 323 | 91 | |
AF081194.1 | - | 2 - 3 | 230 | 91 | |
AK092882.1 | - | 1 - 2 | 993 | 91 | |
AK124023.1 | - | 1 - 2 | 1793 | 91 | |
AK314637.1 | - | 1 - 2 | 208 | 91 | |
BC110306.1 | - | 2 - 3 | 231 | 91 | |
BC117151.1 | - | 2 - 3 | 321 | 91 | |
DA286186.1 | - | 2 - 3 | 317 | 91 | |
DA529151.1 | - | 2 - 3 | 297 | 91 | |
DB498812.1 | - | 2 - 3 | 257 | 91 | |
U78170.1 | - | 2 - 3 | 230 | 91 | |
Y12336.1 | - | 2 - 3 | 323 | 91 |