Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FTO
Entrez Gene ID
79068
Gene Name
fat mass and obesity associated
Gene Aliases
ALKBH9, BMIQ14, GDFD
Chromosome Location
Chr.16: 53703963 - 54114467 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080432.2 NP_001073901.1 5 - 6 1197 97
XM_011523313.2 XP_011521615.1 5 - 6 1191 97
XM_011523314.2 XP_011521616.1 5 - 6 1191 97
XM_011523315.2 XP_011521617.1 5 - 6 1191 97
XM_011523316.2 XP_011521618.1 5 - 6 1191 97
XM_017023654.1 XP_016879143.1 5 - 6 1191 97
XM_017023655.1 XP_016879144.1 5 - 6 1161 97
XM_017023656.1 XP_016879145.1 5 - 6 1161 97
XM_017023657.1 XP_016879146.1 5 - 6 1161 97
GenBank mRNA AK096554.1 - 5 - 6 1004 97
AK295196.1 - 4 - 5 350 97

View Full Product Details