The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005755.2 | NP_005746.2 | 1 - 2 | 76 | 138 |
XM_011527619.2 | XP_011525921.1 | 2 - 3 | 325 | 138 | |
GenBank mRNA | BC015364.1 | - | 1 - 2 | 121 | 138 |
BC046112.1 | - | 1 - 2 | 75 | 138 | |
L08187.1 | - | 1 - 2 | 81 | 138 |