Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPP8
Entrez Gene ID
54878
Gene Name
dipeptidyl peptidase 8
Gene Aliases
DP8, DPRP-1, DPRP1, MST097, MSTP097, MSTP135, MSTP141
Chromosome Location
Chr.15: 65442463 - 65517713 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320875.1 NP_001307804.1 17 - 18 2499 119
NM_001320876.1 NP_001307805.1 16 - 17 2468 119
NM_130434.4 NP_569118.1 16 - 17 2363 119
NM_197960.3 NP_932064.1 17 - 18 2404 119
XM_005254505.1 XP_005254562.1 8 - 9 1045 119
XM_011521730.1 XP_011520032.1 15 - 16 3623 119
XM_011521731.1 XP_011520033.1 15 - 16 3578 119
XM_011521732.2 XP_011520034.1 16 - 17 3752 119
XM_011521733.1 XP_011520035.1 15 - 16 2170 119
XM_017022373.1 XP_016877862.1 16 - 17 3752 119
XM_017022375.1 XP_016877864.1 15 - 16 2215 119
XM_017022377.1 XP_016877866.1 14 - 15 2041 119
XM_017022380.1 XP_016877869.1 15 - 16 2170 119
XM_017022382.1 XP_016877871.1 9 - 10 1172 119
XM_017022383.1 XP_016877872.1 9 - 10 1144 119
GenBank mRNA AF173382.1 - 4 - 5 494 120
AF175225.1 - 4 - 5 498 119
AF176779.1 - 4 - 5 495 119
AF221634.1 - 16 - 17 2337 119
AF221637.1 - 7 - 8 877 119
AY172659.1 - 15 - 16 2131 119
AY354202.1 - 17 - 18 2332 119

View Full Product Details