The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320875.1 | NP_001307804.1 | 8 - 9 | 1331 | 111 |
NM_001320876.1 | NP_001307805.1 | 7 - 8 | 1300 | 111 | |
NM_017743.5 | NP_060213.2 | 7 - 8 | 1195 | 111 | |
NM_130434.4 | NP_569118.1 | 7 - 8 | 1195 | 111 | |
NM_197960.3 | NP_932064.1 | 8 - 9 | 1236 | 111 | |
NM_197961.3 | NP_932065.1 | 8 - 9 | 1331 | 111 | |
NR_135485.1 | 7 - 8 | 1195 | 111 | ||
NR_135486.1 | 8 - 9 | 1236 | 111 | ||
XM_011521731.1 | XP_011520033.1 | 6 - 7 | 2410 | 111 | |
XM_011521732.2 | XP_011520034.1 | 7 - 8 | 2584 | 111 | |
XM_011521733.1 | XP_011520035.1 | 6 - 7 | 1002 | 111 | |
XM_011521734.1 | XP_011520036.1 | 7 - 8 | 2584 | 111 | |
XM_017022373.1 | XP_016877862.1 | 7 - 8 | 2584 | 111 | |
XM_017022374.1 | XP_016877863.1 | 7 - 8 | 2584 | 111 | |
XM_017022376.1 | XP_016877865.1 | 7 - 8 | 1176 | 111 | |
XM_017022378.1 | XP_016877867.1 | 7 - 8 | 2584 | 111 | |
XM_017022379.1 | XP_016877868.1 | 6 - 7 | 1002 | 111 | |
XM_017022380.1 | XP_016877869.1 | 6 - 7 | 1002 | 111 | |
GenBank mRNA | AF221634.1 | - | 7 - 8 | 1169 | 111 |
AK000290.1 | - | 7 - 8 | 1118 | 111 | |
AK027826.1 | - | 3 - 4 | 374 | 111 | |
AK291057.1 | - | 8 - 9 | 1306 | 111 | |
AK296915.1 | - | 7 - 8 | 111 | ||
AK302638.1 | - | 8 - 9 | 1172 | 111 | |
AY172659.1 | - | 6 - 7 | 963 | 111 | |
AY354202.1 | - | 8 - 9 | 1164 | 111 | |
BC030688.2 | - | 8 - 9 | 1322 | 111 |