The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000293.2 | NP_000284.1 | 4 - 5 | 459 | 116 |
NM_001031835.2 | NP_001027005.1 | 5 - 6 | 571 | 116 | |
XM_005255983.4 | XP_005256040.1 | 4 - 5 | 422 | 116 | |
XM_005255984.4 | XP_005256041.1 | 5 - 6 | 722 | 116 | |
XM_017023283.1 | XP_016878772.1 | 5 - 6 | 544 | 116 | |
XM_017023284.1 | XP_016878773.1 | 4 - 5 | 425 | 116 | |
GenBank mRNA | AK309226.1 | - | 4 - 5 | 422 | 116 |
BC033657.1 | - | 5 - 6 | 550 | 116 | |
CR936710.1 | - | 5 - 6 | 524 | 116 | |
X84908.1 | - | 4 - 5 | 431 | 116 |