The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032360.3 | NP_115736.1 | 4 - 5 | 1081 | 94 |
GenBank mRNA | AK314112.1 | - | 4 - 5 | 1056 | 94 |
AY606133.1 | - | 4 - 5 | 495 | 95 | |
BC006505.2 | - | 4 - 5 | 538 | 94 |