The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017433.4 | NP_059129.3 | 16 - 17 | 2027 | 159 |
XM_011519498.2 | XP_011517800.1 | 16 - 17 | 1871 | 159 | |
XM_011519499.1 | XP_011517801.1 | 16 - 17 | 1848 | 159 | |
XM_011519500.2 | XP_011517802.1 | 17 - 18 | 1917 | 159 | |
XM_011519502.1 | XP_011517804.1 | 16 - 17 | 1871 | 159 | |
XM_011519503.1 | XP_011517805.1 | 16 - 17 | 1871 | 159 | |
XM_011519504.1 | XP_011517806.1 | 16 - 17 | 1871 | 159 | |
XM_011519505.1 | XP_011517807.1 | 16 - 17 | 1871 | 159 | |
XM_011519506.2 | XP_011517808.1 | 16 - 17 | 1871 | 159 | |
XM_011519508.2 | XP_011517810.1 | 16 - 17 | 1871 | 159 | |
XM_011519510.1 | XP_011517812.1 | 16 - 17 | 1871 | 159 | |
XM_011519511.2 | XP_011517813.1 | 16 - 17 | 1871 | 159 | |
GenBank mRNA | AF229172.1 | - | 16 - 17 | 1845 | 159 |
AK310494.1 | - | 16 - 17 | 1871 | 159 | |
AY101367.1 | - | 16 - 17 | 1845 | 159 |